missing translation for 'onlineSavingsMsg'
Learn More

Tris[1-phenylisoquinoline-C2,N]iridium(III) (purified by sublimation) 99.0+%, TCI America™

Catalog No. t2685100mg
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T2685100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T2685100MG Supplier TCI America Supplier No. T2685100MG

Chemical Identifiers

CAS 435293-93-9
Molecular Formula C45H30IrN3
Molecular Weight (g/mol) 804.973
MDL Number MFCD09842770
InChI Key NDBCGHNTWCYIIU-UHFFFAOYSA-N
Synonym Ir(piq)3
PubChem CID 53384374
IUPAC Name iridium(3+);1-phenylisoquinoline
SMILES C1=CC=C([C-]=C1)C2=NC=CC3=CC=CC=C32.C1=CC=C([C-]=C1)C2=NC=CC3=CC=CC=C32.C1=CC=C([C-]=C1)C2=NC=CC3=CC=CC=C32.[Ir+3]

Specifications

Melting Point 454°C
Color Red-Yellow
Quantity 100 mg
Formula Weight 804.97
Percent Purity ≥99.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Tris[1-phenylisoquinoline-C2,N]iridium(III) (purified by sublimation)