accessibility menu, dialog, popup

UserName

CNCNCNIr

CAS RN 435293-93-9

IUPAC Name: iridium(3+) tris(2-(isoquinolin-1-yl)benzen-1-ide)
Synonyms: iridium(3+) tris(2-(isoquinolin-1-yl)benzen-1-ide)
Molecular Weight (g/mol): 804.97
Molecular Formula: C45H30IrN3
InChi Key: NDBCGHNTWCYIIU-UHFFFAOYSA-N
SMILES: [Ir+3].C1=CC=C([C-]=C1)C1=C2C=CC=CC2=CC=N1.C1=CC=C([C-]=C1)C1=C2C=CC=CC2=CC=N1.C1=CC=C([C-]=C1)C1=C2C=CC=CC2=CC=N1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results