missing translation for 'onlineSavingsMsg'
Learn More

Tris(1,3-dichloro-2-propyl) Phosphate 95.0+%, TCI America™

Catalog No. p-7134524
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P026925G 25 g
P0269500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P026925G Supplier TCI America Supplier No. P026925G
Only null left

Chemical Identifiers

CAS 13674-87-8
Molecular Formula C9H15Cl6O4P
Molecular Weight (g/mol) 430.889
MDL Number MFCD00083121
InChI Key ASLWPAWFJZFCKF-UHFFFAOYSA-N
Synonym Phosphoric Acid Tris(1,3-dichloro-2-propyl) Ester
PubChem CID 26177
IUPAC Name tris(1,3-dichloropropan-2-yl) phosphate
SMILES C(C(CCl)OP(=O)(OC(CCl)CCl)OC(CCl)CCl)Cl

Specifications

Color Yellow
Boiling Point 252°C
Quantity 25 g
UN Number 3082
Formula Weight 430.89
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Tris(1,3-dichloro-2-propyl) Phosphate
EINECSNumber (2)-1914
RTECSNumber UB1473000
TSCA Yes