missing translation for 'onlineSavingsMsg'
Learn More

Triruthenium Dodecacarbonyl, TCI America™

Catalog No. T21811G
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T21811G 1 g
T2181100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T21811G Supplier TCI America Supplier No. T21811G
Only null left

Chemical Identifiers

CAS 15243-33-1
Molecular Formula C12O12Ru3
Molecular Weight (g/mol) 639.33
MDL Number MFCD00011209
InChI Key NQZFAUXPNWSLBI-UHFFFAOYSA-N
Synonym Ruthenium Carbonyl
IUPAC Name cyclotriruthenium; dodecakis(methanidylidyneoxidanium)
SMILES [C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[Ru]1[Ru][Ru]1

Specifications

Melting Point 150°C
Quantity 1 g
UN Number 3178
Formula Weight 639.33
Physical Form Crystalline Powder
Chemical Name or Material Triruthenium Dodecacarbonyl