accessibility menu, dialog, popup

UserName

CAS RN 15243-33-1

RuRuRuCCCCCCCCCOOOOOOOOOOCCOCO

CAS RN 15243-33-1

IUPAC Name: cyclotriruthenium; dodecakis(methanidylidyneoxidanium)
Synonyms: dodecakis(carbon monoxide); cyclotriruthenium
Molecular Weight (g/mol): 639.33
Molecular Formula: C12O12Ru3
InChi Key: NQZFAUXPNWSLBI-UHFFFAOYSA-N
SMILES: [C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[Ru]1[Ru][Ru]1
Physical Form 
  • (2)
  • (2)
  • (2)
Quantity 
  • (3)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)