missing translation for 'onlineSavingsMsg'
Learn More

Triruthenium dodecacarbonyl, 99%

Catalog No. p-3737521
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC224000010 1 g Glass bottle
AC224000050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC224000010 Supplier Thermo Scientific Chemicals Supplier No. 224000010
Only null left

CAS: 15243-33-1 | C12O12Ru3 | 639.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 15243-33-1
Molecular Formula C12O12Ru3
Molecular Weight (g/mol) 639.33
MDL Number MFCD00011209
InChI Key NQZFAUXPNWSLBI-UHFFFAOYSA-N
Synonym Ruthenium carbonyl
IUPAC Name cyclotriruthenium; dodecakis(methanidylidyneoxidanium)
SMILES [C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[Ru]1[Ru][Ru]1

Specifications

Melting Point 150.0°C
Color Orange
Elemental Analysis 98.5% min.
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula Ru3(CO)12
Quantity 1 g
Solubility Information Solubility in water: insoluble.
Formula Weight 639.33
Percent Purity 99%
Physical Form Crystals
Chemical Name or Material Triruthenium dodecacarbonyl
Show More Show Less
EINECSNumber 239-287-4

RUO – Research Use Only