missing translation for 'onlineSavingsMsg'
Learn More

Triptycene-9,10-dicarboxaldehyde 96.0+%, TCI America™

Catalog No. p-7137604
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3146200MG 200 mg
T31461G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T3146200MG Supplier TCI America Supplier No. T3146200MG
Only null left

Chemical Identifiers

CAS 467429-73-8
Molecular Formula C22H14O2
Molecular Weight (g/mol) 310.35
MDL Number MFCD29472555
InChI Key HTLPGDCICALQIK-UHFFFAOYSA-N
Synonym 9,10-Diformyltriptycene
PubChem CID 131637027
IUPAC Name pentacyclo[6.6.6.0²,⁷.0⁹,¹⁴.0¹⁵,²⁰]icosa-2,4,6,9,11,13,15,17,19-nonaene-1,8-dicarbaldehyde
SMILES O=CC12C3=CC=CC=C3C(C=O)(C3=CC=CC=C13)C1=CC=CC=C21

Specifications

Color Yellow
Quantity 200 mg
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Triptycene-9,10-dicarboxaldehyde