accessibility menu, dialog, popup

UserName

CAS RN 467429-73-8

OO

CAS RN 467429-73-8

IUPAC Name: pentacyclo[6.6.6.0²,⁷.0⁹,¹⁴.0¹⁵,²⁰]icosa-2,4,6,9,11,13,15,17,19-nonaene-1,8-dicarbaldehyde
Synonyms: pentacyclo[6.6.6.0²,⁷.0⁹,¹⁴.0¹⁵,²⁰]icosa-2,4,6,9,11,13,15,17,19-nonaene-1,8-dicarbaldehyde
Molecular Weight (g/mol): 310.35
Molecular Formula: C22H14O2
InChi Key: HTLPGDCICALQIK-UHFFFAOYSA-N
SMILES: O=CC12C3=CC=CC=C3C(C=O)(C3=CC=CC=C13)C1=CC=CC=C21
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results