Learn More
Tripropylene glycol
Description
Tripropylene glycol is used for textile soaps, lubricants, cutting oils and greases. It acts as an intermediate in the preparation of urethanes. It is also used as a solvent in coatings and inks. It is utilized as varnish removers, emulsifiers and hydraulic fluids. As an additive, it is used for carburetor fuels. It is involved as a cooling media as well as an active ingredient of plasticizers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Density | 1.018 g/mL |
| Boiling Point | 268°C |
| Odor | Odorless |
| Refractive Index | 1.4443 |
| Quantity | 250 mL |
| Solubility Information | Miscible with water,methanol and ether. |
| Formula Weight | 192.25 |
| Physical Form | Liquid |
| Chemical Name or Material | Tripropylene glycol |
Safety and Handling
| EINECSNumber | 246-466-0 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only