accessibility menu, dialog, popup

UserName

H3COHOCH3OOHCH3

CAS RN 24800-44-0

IUPAC Name: 1-{[1-(1-hydroxypropoxy)propan-2-yl]oxy}propan-1-ol
Synonyms: tripropylene glycol
Molecular Weight (g/mol): 192.26
Molecular Formula: C9H20O4
InChi Key: SNAJJYBUNRAGEF-UHFFFAOYNA-N
SMILES: CCC(O)OCC(C)OC(O)CC
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (2)
  • (3)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (6)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (2)
  • (3)
  • (5)
  • (3)