missing translation for 'onlineSavingsMsg'
Learn More

Tripropylene Glycol (mixture of isomers) 98.0+%, TCI America™

Catalog No. T0523500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0523500ML 500 mL
T052325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T0523500ML Supplier TCI America Supplier No. T0523500ML

Chemical Identifiers

CAS 24800-44-0
Molecular Formula C9H20O4
MDL Number MFCD00014405

Specifications

Color Yellow
Quantity 500 mL
Formula Weight 192.26
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Tripropylene Glycol (mixture of isomers)
EINECSNumber (2)-0430
RTECSNumber YK6825000
TSCA Yes