Learn More
Chemical Identifiers
| CAS | 621-76-1 |
|---|---|
| Molecular Formula | C10H22O3 |
| Molecular Weight (g/mol) | 190.29 |
| MDL Number | MFCD00015214 |
| InChI Key | RWNXXQFJBALKAX-UHFFFAOYSA-N |
| Synonym | tripropyl orthoformate, tri-n-propyl orthoformate, orthoformic acid tripropyl ester, 1-dipropoxymethoxy propane, unii-z8f0df6af9, propane, 1,1',1-methylidynetris oxy tris, z8f0df6af9, tripropoxymethane, tripropylorthoformate |
| PubChem CID | 69311 |
| IUPAC Name | 1-(dipropoxymethoxy)propane |
| SMILES | CCCOC(OCCC)OCCC |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC369290250
|
N/A | 25mL | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Alkylation reagentChemical Identifiers
| C10H22O3 | |
| MFCD00015214 | |
| tripropyl orthoformate, tri-n-propyl orthoformate, orthoformic acid tripropyl ester, 1-dipropoxymethoxy propane, unii-z8f0df6af9, propane, 1,1',1-methylidynetris oxy tris, z8f0df6af9, tripropoxymethane, tripropylorthoformate | |
| 1-(dipropoxymethoxy)propane |
Specifications
| 621-76-1 | |
| 106°C to 108°C (40.0mmHg) | |
| Authentic | |
| C10H22O3 | |
| CH(OCH2CH2CH3)3 | |
| 02, IV, 27 | |
| tripropyl orthoformate, tri-n-propyl orthoformate, orthoformic acid tripropyl ester, 1-dipropoxymethoxy propane, unii-z8f0df6af9, propane, 1,1',1-methylidynetris oxy tris, z8f0df6af9, tripropoxymethane, tripropylorthoformate | |
| CCCOC(OCCC)OCCC | |
| 190.29 | |
| 190.29 | |
| Tripropyl orthoformate |
| 0.8790g/mL | |
| 72°C | |
| 97.5% min. (GC) | |
| 1.4060 to 1.408 | |
| MFCD00015214 | |
| 0.879 | |
| RWNXXQFJBALKAX-UHFFFAOYSA-N | |
| 1-(dipropoxymethoxy)propane | |
| 69311 | |
| 98% |
Safety and Handling
GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 210-704-1
RUO – Research Use Only