accessibility menu, dialog, popup

UserName

CH3OOCH3OCH3

CAS RN 621-76-1

IUPAC Name: 1-(dipropoxymethoxy)propane
Synonyms: 1-dipropoxymethoxy-propane
Molecular Weight (g/mol): 190.28
Molecular Formula: C10H22O3
InChi Key: RWNXXQFJBALKAX-UHFFFAOYSA-N
SMILES: CCCOC(OCCC)OCCC
Boiling Point 
  • (3)
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (4)
  • (1)
Percent Purity 
  • (1)
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (4)
  • (3)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)