missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tripropyl Orthoformate 98.0+%, TCI America™
$88.56 - $88.56
Chemical Identifiers
| CAS | 621-76-1 |
|---|---|
| Molecular Formula | C10H22O3 |
| Molecular Weight (g/mol) | 190.283 |
| MDL Number | MFCD00015214 |
| InChI Key | RWNXXQFJBALKAX-UHFFFAOYSA-N |
| Synonym | tripropyl orthoformate, tri-n-propyl orthoformate, orthoformic acid tripropyl ester, 1-dipropoxymethoxy propane, unii-z8f0df6af9, propane, 1,1',1-methylidynetris oxy tris, z8f0df6af9, tripropoxymethane, tripropylorthoformate |
| PubChem CID | 69311 |
| IUPAC Name | 1-(dipropoxymethoxy)propane |
| SMILES | CCCOC(OCCC)OCCC |
Chemical Identifiers
| 621-76-1 | |
| 190.283 | |
| RWNXXQFJBALKAX-UHFFFAOYSA-N | |
| 69311 | |
| CCCOC(OCCC)OCCC |
| C10H22O3 | |
| MFCD00015214 | |
| tripropyl orthoformate, tri-n-propyl orthoformate, orthoformic acid tripropyl ester, 1-dipropoxymethoxy propane, unii-z8f0df6af9, propane, 1,1',1-methylidynetris oxy tris, z8f0df6af9, tripropoxymethane, tripropylorthoformate | |
| 1-(dipropoxymethoxy)propane |
Specifications
| 621-76-1 | |
| 198°C | |
| MFCD00015214 | |
| tripropyl orthoformate, tri-n-propyl orthoformate, orthoformic acid tripropyl ester, 1-dipropoxymethoxy propane, unii-z8f0df6af9, propane, 1,1',1-methylidynetris oxy tris, z8f0df6af9, tripropoxymethane, tripropylorthoformate | |
| CCCOC(OCCC)OCCC | |
| 190.283 | |
| 190.28 | |
| Liquid |
| Yellow | |
| C10H22O3 | |
| 25 mL | |
| RWNXXQFJBALKAX-UHFFFAOYSA-N | |
| 1-(dipropoxymethoxy)propane | |
| 69311 | |
| ≥98.0% (GC) | |
| Tripropyl Orthoformate |
Safety and Handling
EINECSNumber : (2)-3932
TSCA : Yes