missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tripropyl Borate 99.0+%, TCI America™
$83.89 - $561.08
Chemical Identifiers
| CAS | 688-71-1 |
|---|---|
| Molecular Formula | C9H21BO3 |
| Molecular Weight (g/mol) | 188.074 |
| MDL Number | MFCD00009366 |
| InChI Key | LTEHWCSSIHAVOQ-UHFFFAOYSA-N |
| Synonym | tri-n-propyl borate, tripropoxyborane, boron propoxide, propyl orthoborate, tris propoxy borane, boric acid h3bo3 , tripropyl ester, tripropyl orthoborate, boric acid, tripropyl ester, propyl borate, pro 3b, boric acid tri-n-propyl ester |
| PubChem CID | 12711 |
| IUPAC Name | tripropyl borate |
| SMILES | B(OCCC)(OCCC)OCCC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
B013425ML
|
TCI America
B013425ML |
25 mL |
Each for $83.89
|
|
|||||
|
B0134500ML
|
TCI America
B0134500ML |
500 mL |
Each for $561.08
|
|
|||||
Chemical Identifiers
| 688-71-1 | |
| 188.074 | |
| LTEHWCSSIHAVOQ-UHFFFAOYSA-N | |
| 12711 | |
| B(OCCC)(OCCC)OCCC |
| C9H21BO3 | |
| MFCD00009366 | |
| tri-n-propyl borate, tripropoxyborane, boron propoxide, propyl orthoborate, tris propoxy borane, boric acid h3bo3 , tripropyl ester, tripropyl orthoborate, boric acid, tripropyl ester, propyl borate, pro 3b, boric acid tri-n-propyl ester | |
| tripropyl borate |
Specifications
| 688-71-1 | |
| 180°C | |
| MFCD00009366 | |
| 1993 | |
| LTEHWCSSIHAVOQ-UHFFFAOYSA-N | |
| tripropyl borate | |
| 12711 | |
| ≥99.0% (T) | |
| Tripropyl Borate |
| Colorless | |
| C9H21BO3 | |
| 25 mL | |
| tri-n-propyl borate, tripropoxyborane, boron propoxide, propyl orthoborate, tris propoxy borane, boric acid h3bo3 , tripropyl ester, tripropyl orthoborate, boric acid, tripropyl ester, propyl borate, pro 3b, boric acid tri-n-propyl ester | |
| B(OCCC)(OCCC)OCCC | |
| 188.074 | |
| 188.07 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-2101
RTECSNumber : ED5785000
TSCA : Yes