CAS RN 688-71-1
IUPAC Name:
tripropyl borate
Synonyms:
boric acid (H3BO3), tripropyl ester
Molecular Weight (g/mol):
188.07
Molecular Formula:
C9H21BO3
InChi Key:
LTEHWCSSIHAVOQ-UHFFFAOYSA-N
SMILES:
CCCOB(OCCC)OCCC
1
–
1
of
1
results
1
Tripropyl Borate 99.0+%, TCI America™
CAS: 688-71-1 Molecular Formula: C9H21BO3 Molecular Weight (g/mol): 188.074 MDL Number: MFCD00009366 InChI Key: LTEHWCSSIHAVOQ-UHFFFAOYSA-N Synonym: tri-n-propyl borate,tripropoxyborane,boron propoxide,propyl orthoborate,tris propoxy borane,boric acid h3bo3 , tripropyl ester,tripropyl orthoborate,boric acid, tripropyl ester,propyl borate, pro 3b,boric acid tri-n-propyl ester PubChem CID: 12711 IUPAC Name: tripropyl borate SMILES: B(OCCC)(OCCC)OCCC