missing translation for 'onlineSavingsMsg'
Learn More

Tripropyl Borate 99.0+%, TCI America™

Catalog No. B0134500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0134500ML 500 mL
B013425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0134500ML Supplier TCI America Supplier No. B0134500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 688-71-1
Molecular Formula C9H21BO3
Molecular Weight (g/mol) 188.074
MDL Number MFCD00009366
InChI Key LTEHWCSSIHAVOQ-UHFFFAOYSA-N
Synonym tri-n-propyl borate, tripropoxyborane, boron propoxide, propyl orthoborate, tris propoxy borane, boric acid h3bo3 , tripropyl ester, tripropyl orthoborate, boric acid, tripropyl ester, propyl borate, pro 3b, boric acid tri-n-propyl ester
PubChem CID 12711
IUPAC Name tripropyl borate
SMILES B(OCCC)(OCCC)OCCC

Specifications

Color Colorless
Boiling Point 180°C
Quantity 500 mL
UN Number 1993
Formula Weight 188.07
Percent Purity ≥99.0% (T)
Physical Form Liquid
Chemical Name or Material Tripropyl Borate
EINECSNumber (2)-2101
RTECSNumber ED5785000
TSCA Yes