missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tripropionin 95.0+%, TCI America™
$36.07 - $161.38
Chemical Identifiers
| CAS | 139-45-7 |
|---|---|
| Molecular Formula | C12H20O6 |
| Molecular Weight (g/mol) | 260.286 |
| MDL Number | MFCD00027005 |
| InChI Key | YZWRNSARCRTXDS-UHFFFAOYSA-N |
| Synonym | tripropionin, glycerol tripropionate, glyceryl tripropionate, tripropionine, glyceryl tripropanoate, propionin, tri, glycerine tripropionate, tripropionylglycerol, 1,2,3-propanetriol, tripropanoate, propionic acid triglyceride |
| PubChem CID | 8763 |
| IUPAC Name | 2,3-di(propanoyloxy)propyl propanoate |
| SMILES | CCC(=O)OCC(COC(=O)CC)OC(=O)CC |
Chemical Identifiers
| C12H20O6 | |
| MFCD00027005 | |
| tripropionin, glycerol tripropionate, glyceryl tripropionate, tripropionine, glyceryl tripropanoate, propionin, tri, glycerine tripropionate, tripropionylglycerol, 1,2,3-propanetriol, tripropanoate, propionic acid triglyceride | |
| 2,3-di(propanoyloxy)propyl propanoate |
Specifications
| 139-45-7 | |
| 176°C | |
| MFCD00027005 | |
| YZWRNSARCRTXDS-UHFFFAOYSA-N | |
| 2,3-di(propanoyloxy)propyl propanoate | |
| 8763 | |
| ≥95.0% (GC) | |
| Tripropionin |
| Colorless | |
| C12H20O6 | |
| tripropionin, glycerol tripropionate, glyceryl tripropionate, tripropionine, glyceryl tripropanoate, propionin, tri, glycerine tripropionate, tripropionylglycerol, 1,2,3-propanetriol, tripropanoate, propionic acid triglyceride | |
| CCC(=O)OCC(COC(=O)CC)OC(=O)CC | |
| 260.286 | |
| 260.29 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0666
RTECSNumber : YK6000000
TSCA : Yes