missing translation for 'onlineSavingsMsg'
Learn More

Tripropionin 95.0+%, TCI America™

Catalog No. G009025G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G009025G 25 g
G0090500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G009025G Supplier TCI America Supplier No. G009025G

Chemical Identifiers

CAS 139-45-7
Molecular Formula C12H20O6
Molecular Weight (g/mol) 260.286
MDL Number MFCD00027005
InChI Key YZWRNSARCRTXDS-UHFFFAOYSA-N
Synonym tripropionin, glycerol tripropionate, glyceryl tripropionate, tripropionine, glyceryl tripropanoate, propionin, tri, glycerine tripropionate, tripropionylglycerol, 1,2,3-propanetriol, tripropanoate, propionic acid triglyceride
PubChem CID 8763
IUPAC Name 2,3-di(propanoyloxy)propyl propanoate
SMILES CCC(=O)OCC(COC(=O)CC)OC(=O)CC

Specifications

Color Colorless
Boiling Point 176°C
Quantity 25 g
Formula Weight 260.29
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Tripropionin
EINECSNumber (2)-0666
RTECSNumber YK6000000
TSCA Yes