missing translation for 'onlineSavingsMsg'
Learn More

Tripropionin 95.0+%, TCI America™

Supplier:  TCI America G0090500G

 View more versions of this product

Catalog No. G0090500G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Tripropionin
Colorless
C12H20O6
500 g
YZWRNSARCRTXDS-UHFFFAOYSA-N
2,3-di(propanoyloxy)propyl propanoate
8763
≥95.0% (GC)
139-45-7
176°C
MFCD00027005
tripropionin, glycerol tripropionate, glyceryl tripropionate, tripropionine, glyceryl tripropanoate, propionin, tri, glycerine tripropionate, tripropionylglycerol, 1,2,3-propanetriol, tripropanoate, propionic acid triglyceride
CCC(=O)OCC(COC(=O)CC)OC(=O)CC
260.286
260.29
Liquid

Chemical Identifiers

139-45-7
260.286
YZWRNSARCRTXDS-UHFFFAOYSA-N
8763
CCC(=O)OCC(COC(=O)CC)OC(=O)CC
C12H20O6
MFCD00027005
tripropionin, glycerol tripropionate, glyceryl tripropionate, tripropionine, glyceryl tripropanoate, propionin, tri, glycerine tripropionate, tripropionylglycerol, 1,2,3-propanetriol, tripropanoate, propionic acid triglyceride
2,3-di(propanoyloxy)propyl propanoate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0666

RTECSNumber : YK6000000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions