Learn More
TrippNT Step Shelves
Description
Step shelves create storage space above protrusions on the benchtop like gas outlets and other lab supplies. Cords and wires fit through space at back.
Specifications
Specifications
| Product Type | Step-Shelf |
| Height (Metric) | 25.4 cm |
| Height (English) | 10 in. |
| Size | Medium |