missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TrippNT Hood or Wall Mount Pipette Storage Bin
Specifications
Specifications
| Color | White |
| For Use With (Equipment) | Mount with magnet or use free-standing |
| Product Type | Wall or Hood Pipet Bin |