Learn More
TrippNT Hood Shelves
Catalog No. 50856169
Encompass
Change view
Click to view available options
Size:
Small
Medium
Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
50856169 Large
50856168 Medium
50856167 Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 50856169 Supplier TrippNT Supplier No. TRP 50217
Allow air to flow. Keep supplies off hood floor for safety. Keeps fume hood area clear for working, preventing spills and accidents.
Specifications
Product Type Hood Shelf
Size Large