Learn More
Greiner Bio-One Triple-Packed Serological Pipette
Packed in three individual peel bags for sterile transfer of products.
Supplier:  Greiner Bio-One 607160TRI
 View more versions of this product
Catalog No. 07000992
Only null left
Explore available promotions
Description
Description
  • Made from polystyrene
  • Free of heavy metal
  • Not autoclavable
  • Clear graduated pipette
  • Sterile
Specifications
Specifications
± 0.2%
Polystyrene
Yes
Triple packed
10 mL
Includes yellow code ring, cotton plug
Sterile
1/10 mL
Clear
Pipette
Product Suggestions
Product Suggestions