missing translation for 'onlineSavingsMsg'
Learn More

Triphosgene 98.0+%, TCI America™

Catalog No. p-7136792
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T146725G 25 g
T1467250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T146725G Supplier TCI America Supplier No. T146725G
Only null left

Chemical Identifiers

CAS 32315-10-9
Molecular Formula C3Cl6O3
Molecular Weight (g/mol) 296.73
MDL Number MFCD00062848
InChI Key UCPYLLCMEDAXFR-UHFFFAOYSA-N
Synonym triphosgene, bis trichloromethyl carbonate, methanol, trichloro-, carbonate 2:1, ditrichloromethyl carbonate, triphosgene bis-trichloromethyl carbonate, methanol, 1,1,1-trichloro-, 1,1'-carbonate, triphosgen, tri phosgene, tri-phosgene
PubChem CID 94429
IUPAC Name bis(trichloromethyl) carbonate
SMILES C(=O)(OC(Cl)(Cl)Cl)OC(Cl)(Cl)Cl

Specifications

Melting Point 80°C
Color White-Yellow
Boiling Point 206°C
Quantity 25 g
UN Number 2928
Formula Weight 296.73
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Triphosgene
TSCA Yes
Recommended Storage Refrigerator