missing translation for 'onlineSavingsMsg'
Learn More

Triphosgene, 99%

Catalog No. p-4510336
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC258950050 5 g Glass bottle
AC258950100 10 g Glass bottle
AC258950500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC258950050 Supplier Thermo Scientific Chemicals Supplier No. 258950050
Only null left

CAS: 32315-10-9 | C3Cl6O3 | 296.75 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent, C-X bond formation reagent

Chemical Identifiers

CAS 32315-10-9
Molecular Formula C3Cl6O3
Molecular Weight (g/mol) 296.75
InChI Key UCPYLLCMEDAXFR-UHFFFAOYSA-N
Synonym triphosgene, bis trichloromethyl carbonate, methanol, trichloro-, carbonate 2:1, ditrichloromethyl carbonate, triphosgene bis-trichloromethyl carbonate, methanol, 1,1,1-trichloro-, 1,1'-carbonate, triphosgen, tri phosgene, tri-phosgene
PubChem CID 94429
IUPAC Name bis(trichloromethyl) carbonate
SMILES C(=O)(OC(Cl)(Cl)Cl)OC(Cl)(Cl)Cl

Specifications

Melting Point 77.0°C to 81.0°C
Color White
Density 1.7800g/mL
Boiling Point 203.0°C to 206.0°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min
Packaging Glass bottle
Linear Formula C(Cl3)OC(O)OC(Cl)3
Quantity 5 g
Fieser 17,127
Merck Index 15,9921
Specific Gravity 1.78
Solubility Information Solubility in water: practically insoluble, may form phosgene. Other solubilities: 588g/L ch2cl2 (20°C), soluble in alcohol, benzene, tetrachloromethane, and ether
Formula Weight 296.75
Percent Purity 99%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material Triphosgene
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Fatal if inhaled.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 250-986-3

RUO – Research Use Only