missing translation for 'onlineSavingsMsg'
Learn More

Triphenyltetrazolium Chloride, 93%, Spectrum™ Chemical

Catalog No. 18614573
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18614573 5 g Amber Glass Bottle
18614574 25 g Amber Glass Bottle
18614575 100 g Amber Glass Bottle
18614576 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18614573 Supplier Spectrum Chemical Mfg Cor Supplier No. T11245GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 298-96-4

Specifications

CAS Max % 1
Loss on Drying 0.03
Packaging Amber Glass Bottle
Quantity 5 g
Percent Purity 0.93
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.