missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triphenylsilanol 98.0+%, TCI America™
$150.98 - $851.87
Chemical Identifiers
| CAS | 791-31-1 |
|---|---|
| Molecular Formula | C18H16OSi |
| Molecular Weight (g/mol) | 276.41 |
| MDL Number | MFCD00002102 |
| InChI Key | NLSXASIDNWDYMI-UHFFFAOYSA-N |
| Synonym | triphenylsilanol, silanol, triphenyl, triphenylhydroxysilane, silane, hydroxytriphenyl, unii-azc8o4ttx4, silanol, 1,1,1-triphenyl, hydroxy triphenyl silane, si oh ph3, azc8o4ttx4 |
| PubChem CID | 69925 |
| ChEBI | CHEBI:35035 |
| IUPAC Name | hydroxy(triphenyl)silane |
| SMILES | C1=CC=C(C=C1)[Si](C2=CC=CC=C2)(C3=CC=CC=C3)O |
Chemical Identifiers
| 791-31-1 | |
| 276.41 | |
| NLSXASIDNWDYMI-UHFFFAOYSA-N | |
| 69925 | |
| hydroxy(triphenyl)silane |
| C18H16OSi | |
| MFCD00002102 | |
| triphenylsilanol, silanol, triphenyl, triphenylhydroxysilane, silane, hydroxytriphenyl, unii-azc8o4ttx4, silanol, 1,1,1-triphenyl, hydroxy triphenyl silane, si oh ph3, azc8o4ttx4 | |
| CHEBI:35035 | |
| C1=CC=C(C=C1)[Si](C2=CC=CC=C2)(C3=CC=CC=C3)O |
Specifications
| 791-31-1 | |
| White | |
| MFCD00002102 | |
| triphenylsilanol, silanol, triphenyl, triphenylhydroxysilane, silane, hydroxytriphenyl, unii-azc8o4ttx4, silanol, 1,1,1-triphenyl, hydroxy triphenyl silane, si oh ph3, azc8o4ttx4 | |
| C1=CC=C(C=C1)[Si](C2=CC=CC=C2)(C3=CC=CC=C3)O | |
| 276.41 | |
| CHEBI:35035 | |
| ≥98.0% (GC) | |
| Triphenylsilanol |
| 153°C | |
| C18H16OSi | |
| 25 g | |
| NLSXASIDNWDYMI-UHFFFAOYSA-N | |
| hydroxy(triphenyl)silane | |
| 69925 | |
| 276.41 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (3)-2638
RTECSNumber : VV4325500
TSCA : Yes