Learn More
Triphenylsilanol, 98%
CAS: 791-31-1 | C18H16OSi | 276.41 g/mol
Supplier: Thermo Scientific Chemicals A1928422
Description
Triphenylsilanol is used as a water surrogate for regioselective Pd catalyzed allylations. A thick on germanium substrates have been produced by the electron bombardment of an evaporated thin film of triphenylsilanol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTriphenylsilanol is used as a water surrogate for regioselective Pd catalyzed allylations. A thick on germanium substrates have been produced by the electron bombardment of an evaporated thin film of triphenylsilanol.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.
Specifications
| Triphenylsilanol | |
| 791-31-1 | |
| C18H16OSi | |
| 985007 | |
| Insoluble in water. | |
| C1=CC=C(C=C1)[Si](C2=CC=CC=C2)(C3=CC=CC=C3)O | |
| 276.41 | |
| CHEBI:35035 | |
| 98% | |
| 1.17 |
| 149°C to 154°C | |
| 100 g | |
| MFCD00002102 | |
| triphenylsilanol, silanol, triphenyl, triphenylhydroxysilane, silane, hydroxytriphenyl, unii-azc8o4ttx4, silanol, 1,1,1-triphenyl, hydroxy triphenyl silane, si oh ph3, azc8o4ttx4 | |
| NLSXASIDNWDYMI-UHFFFAOYSA-N | |
| hydroxy(triphenyl)silane | |
| 69925 | |
| 276.41 | |
| Moisture sensitive |
Chemical Identifiers
| 791-31-1 | |
| 276.41 | |
| NLSXASIDNWDYMI-UHFFFAOYSA-N | |
| 69925 | |
| hydroxy(triphenyl)silane |
| C18H16OSi | |
| MFCD00002102 | |
| triphenylsilanol, silanol, triphenyl, triphenylhydroxysilane, silane, hydroxytriphenyl, unii-azc8o4ttx4, silanol, 1,1,1-triphenyl, hydroxy triphenyl silane, si oh ph3, azc8o4ttx4 | |
| CHEBI:35035 | |
| C1=CC=C(C=C1)[Si](C2=CC=CC=C2)(C3=CC=CC=C3)O |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 212-339-3
RTECSNumber : VV4325500
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only