missing translation for 'onlineSavingsMsg'
Learn More

Triphenylsilanol, 98%

Catalog No. AAA1928406
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1928406 5 g
AAA1928414 25 g
AAA1928422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1928406 Supplier Thermo Scientific Chemicals Supplier No. A1928406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 791-31-1 | C18H16OSi | 276.41 g/mol

Triphenylsilanol is used as a water surrogate for regioselective Pd catalyzed allylations. A thick on germanium substrates have been produced by the electron bombardment of an evaporated thin film of triphenylsilanol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triphenylsilanol is used as a water surrogate for regioselective Pd catalyzed allylations. A thick on germanium substrates have been produced by the electron bombardment of an evaporated thin film of triphenylsilanol.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.

Chemical Identifiers

CAS 791-31-1
Molecular Formula C18H16OSi
Molecular Weight (g/mol) 276.41
MDL Number MFCD00002102
InChI Key NLSXASIDNWDYMI-UHFFFAOYSA-N
Synonym triphenylsilanol, silanol, triphenyl, triphenylhydroxysilane, silane, hydroxytriphenyl, unii-azc8o4ttx4, silanol, 1,1,1-triphenyl, hydroxy triphenyl silane, si oh ph3, azc8o4ttx4
PubChem CID 69925
ChEBI CHEBI:35035
IUPAC Name hydroxy(triphenyl)silane
SMILES C1=CC=C(C=C1)[Si](C2=CC=CC=C2)(C3=CC=CC=C3)O

Specifications

Melting Point 149°C to 154°C
Quantity 5 g
Sensitivity Moisture sensitive
Density 1.17
Beilstein 985007
Solubility Information Insoluble in water.
Formula Weight 276.41
Percent Purity 98%
Chemical Name or Material Triphenylsilanol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 212-339-3
RTECSNumber VV4325500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only