missing translation for 'onlineSavingsMsg'
Learn More

Triphenylphosphonium bromide, 99%

Catalog No. AC269090250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC269090250 25 g Glass bottle
AC269091000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC269090250 Supplier Thermo Scientific Chemicals Supplier No. 269090250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 6399-81-1 | C18H16BrP | 343.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For C-C Bond Formation (Olefination)

Chemical Identifiers

CAS 6399-81-1
Molecular Formula C18H16BrP
Molecular Weight (g/mol) 343.20
MDL Number MFCD00035107
InChI Key CMSYDJVRTHCWFP-UHFFFAOYSA-N
Synonym triphenylphosphine hydrobromide, triphenylphosphonium bromide, phosphine, triphenyl-, hydrobromide, triphenylphosphane hydrobromide, triphenylphosphine bromine, pubchem13868, ph3p hbr, ph3p . hbr, unappfzuzwmopv-uhfffaoysa-n
PubChem CID 80811
IUPAC Name triphenylphosphane;hydrobromide
SMILES Br.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 196.0°C to 199.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Argentometry)
Packaging Glass bottle
Quantity 25 g
Fieser 15,353
Formula Weight 343.2
Percent Purity 99%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material Triphenylphosphonium bromide, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 229-012-6

RUO – Research Use Only