missing translation for 'onlineSavingsMsg'
Learn More

Triphenylphosphine Sulfide 99.0+%, TCI America™

Catalog No. t1066500g
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T1066500G 500 g
T106625G 25 g
T1066100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T1066500G Supplier TCI America Supplier No. T1066500G
Only null left

Chemical Identifiers

CAS 3878-45-3
Molecular Formula C18H15PS
Molecular Weight (g/mol) 294.35
MDL Number MFCD00004917
InChI Key VYNGFCUGSYEOOZ-UHFFFAOYSA-N
Synonym triphenylphosphine sulfide, phosphine sulfide, triphenyl, triphenylphosphine sulphide, triphenylphosphine monosulfide, triphenylphosphane sulfide, triphenylphosphino-1-thione, triphenylthioxophosphorane, triphenylphosphorus sulfide, wln: spr&r&r, triphenyl-, e5-phosphanethione
PubChem CID 19758
IUPAC Name triphenyl-λ⁵-phosphanethione
SMILES S=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 163°C
Color White
Quantity 500 g
Formula Weight 294.35
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Triphenylphosphine Sulfide
EINECSNumber (3)-2610
RTECSNumber SZ2820000
TSCA No