missing translation for 'onlineSavingsMsg'
Learn More

Triphenylmethylium Tetrafluoroborate 98.0+%, TCI America™

Catalog No. p-7136642
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T11735G 5 g
T117325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T11735G Supplier TCI America Supplier No. T11735G
Only null left

Chemical Identifiers

CAS 341-02-6
Molecular Formula C19H15BF4
Molecular Weight (g/mol) 330.133
MDL Number MFCD00013120
InChI Key VQXBOEYKSVVPSP-UHFFFAOYSA-N
Synonym triphenylcarbenium tetrafluoroborate, trityl tetrafluoroborate, tritylium tetrafluoroborate, triphenylmethylium tetrafluoroborate, diphenylmethylbenzene tetrafluoroborate, trityl fluoroborate, triphenylmethyl fluoroborate, triphenylcarbonium tetrafluoroborate, methylium, triphenyl-, tetrafluoroborate 1-, methylium, triphenyl-, tetrafluoroborate 1-1:1
PubChem CID 2723955
IUPAC Name diphenylmethylbenzene;tetrafluoroborate
SMILES [B-](F)(F)(F)F.C1=CC=C(C=C1)[C+](C2=CC=CC=C2)C3=CC=CC=C3

Specifications

Melting Point 202°C
Quantity 5 g
UN Number 1759
Formula Weight 330.13
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Triphenylmethylium Tetrafluoroborate
TSCA Yes
Recommended Storage Refrigerator