accessibility menu, dialog, popup

UserName

CAS RN 341-02-6

IUPAC Name: tetrafluoroboranuide; triphenylmethylium
Synonyms: triphenylmethylium tetrafluoroborate
Molecular Weight (g/mol): 330.13
Molecular Formula: C19H15BF4
InChi Key: VQXBOEYKSVVPSP-UHFFFAOYSA-N
SMILES: F[B-](F)(F)F.C1=CC=C(C=C1)[C+](C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (5)
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (5)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)