missing translation for 'onlineSavingsMsg'
Learn More

Triphenylmethylamine 95.0+%, TCI America™

Catalog No. T05175G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T05175G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T05175G Supplier TCI America Supplier No. T05175G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5824-40-8
Molecular Formula C19H17N
Molecular Weight (g/mol) 259.352
MDL Number MFCD00008047
InChI Key BZVJOYBTLHNRDW-UHFFFAOYSA-N
Synonym tritylamine, triphenylmethylamine, benzenemethanamine, .alpha.,.alpha.-diphenyl, tritylamin, trityl amine, aminotriphenylmethane, triphenylaminomethane, triphenyl methylamine, aminotrityl, polymer-bound
PubChem CID 138598
IUPAC Name triphenylmethanamine
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)N

Specifications

Melting Point 102°C
Color White-Yellow
Boiling Point 223°C
Quantity 5 g
Formula Weight 259.35
Percent Purity ≥95.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Triphenylmethylamine