missing translation for 'onlineSavingsMsg'
Learn More

Triphenylmethyl mercaptan, 98+%

CAS: 3695-77-0 | C19H16S | 276.397 g/mol

Supplier:  Thermo Scientific Chemicals A1488814

 View more versions of this product

Catalog No. AAA1488814

Only null left
Explore available promotions

Description

Description

Triphenylmethyl mercaptan, is used as a reagent used for the introduction of thiol group. Triphenylmethanethiol is a reagent used for the mild introduction of the mercapto functionality. It is also used as a pharmaceutical intermediate; Chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triphenylmethyl mercaptan, is used as a reagent used for the introduction of thiol group. Triphenylmethanethiol is a reagent used for the mild introduction of the mercapto functionality. It is also used as a pharmaceutical intermediate; Chemical synthesis.

Solubility
It is insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents. Stable under recommended storage conditions.
Specifications

Specifications

Triphenylmethyl mercaptan
103°C to 108°C
C19H16S
25 g
triphenylmethyl mercaptan, trityl mercaptan, tritylthiol, triphenylmethylmercaptan, methanethiol, triphenyl, tritylmercaptan, triphenyl-methanethiol, alpha,alpha-diphenylbenzenemethanethiol, benzenemethanethiol, alpha,alpha-diphenyl, benzenemethanethiol, .alpha.,.alpha.-diphenyl
JQZIKLPHXXBMCA-UHFFFAOYSA-N
triphenylmethanethiol
77281
≥98%
3695-77-0
Stench
MFCD00004854
2113168
It is insoluble in water.
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)S
276.397
276.4

Chemical Identifiers

3695-77-0
276.397
JQZIKLPHXXBMCA-UHFFFAOYSA-N
77281
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)S
C19H16S
MFCD00004854
triphenylmethyl mercaptan, trityl mercaptan, tritylthiol, triphenylmethylmercaptan, methanethiol, triphenyl, tritylmercaptan, triphenyl-methanethiol, alpha,alpha-diphenylbenzenemethanethiol, benzenemethanethiol, alpha,alpha-diphenyl, benzenemethanethiol, .alpha.,.alpha.-diphenyl
triphenylmethanethiol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P271-P304+P340-P312

H332

EINECSNumber : 223-020-3

RTECSNumber : PB5335000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only