missing translation for 'onlineSavingsMsg'
Learn More

Triphenylmethanol, 98%

Catalog No. AAA1036618
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1036618 50 g
AAA1036630 250 g
AAA103660B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1036618 Supplier Thermo Scientific Chemicals Supplier No. A1036618
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 76-84-6 | C19H16O | 260.34 g/mol

Triphenylmethanol is used as a reagent in the research laboratory. It acts as an intermediate in the production of the commercially useful triarylmethane dyes. It is used in the preparation of triphenylmethane. It is also used as an antiproliferative agent. Further, it is used in the preparation of two-electron reduction product of pyrylogen. In addition to this, it reacts with triphenylphosphine oxide to form a 1:1 molecular complex. It serves as a specific clathrate host for methanol and dimethyl sulphoxide and forms clathrate inclusion complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triphenylmethanol is used as a reagent in the research laboratory. It acts as an intermediate in the production of the commercially useful triarylmethane dyes. It is used in the preparation of triphenylmethane. It is also used as an antiproliferative agent. Further, it is used in the preparation of two-electron reduction product of pyrylogen. In addition to this, it reacts with triphenylphosphine oxide to form a 1:1 molecular complex. It serves as a specific clathrate host for methanol and dimethyl sulphoxide and forms clathrate inclusion complexes.

Solubility
Soluble in dioxane, ethanol, water, ether and benzene.

Notes
Incompatible with acids, acid chlorides, acid anhydrides and oxidizing agents.

Chemical Identifiers

CAS 76-84-6
Molecular Formula C19H16O
Molecular Weight (g/mol) 260.34
MDL Number MFCD00004445,MFCD10565638
InChI Key LZTRCELOJRDYMQ-UHFFFAOYSA-N
Synonym triphenylcarbinol, trityl alcohol, tritanol, triphenylmethyl alcohol, triphenyl methanol, triphenyl carbinol, methanol, triphenyl, unii-u97q0ou9kb, triphenyl-methanol, benzenemethanol, .alpha.,.alpha.-diphenyl
PubChem CID 6457
IUPAC Name triphenylmethanol
SMILES OC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 161°C to 164°C
Boiling Point 360°C to 380°C
Assay Percent Range 98%
Quantity 50 g
Beilstein 1460837
Merck Index 14,9739
Solubility Information Soluble in dioxane,ethanol,water,ether and benzene.
Formula Weight 260.34
Percent Purity 98%
Chemical Name or Material Triphenylmethanol
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P362-P312-P321-P405-P403+P233-P501a
EINECSNumber 200-988-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only