missing translation for 'onlineSavingsMsg'
Learn More

Triphenylmethanol, 97%

Catalog No. AC158910500
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC158910500 50 g Plastic bottle
AC158912500 250 g Plastic bottle
AC158910010 1 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC158910500 Supplier Thermo Scientific Chemicals Supplier No. 158910500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 76-84-6 | C19H16O | 260.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 76-84-6
Molecular Formula C19H16O
Molecular Weight (g/mol) 260.34
MDL Number MFCD00004445,MFCD10565638
InChI Key LZTRCELOJRDYMQ-UHFFFAOYSA-N
Synonym triphenylcarbinol, trityl alcohol, tritanol, triphenylmethyl alcohol, triphenyl methanol, triphenyl carbinol, methanol, triphenyl, unii-u97q0ou9kb, triphenyl-methanol, benzenemethanol, .alpha.,.alpha.-diphenyl
PubChem CID 6457
IUPAC Name triphenylmethanol
SMILES OC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 160°C to 164°C
Color White to Yellow
Boiling Point 360°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Plastic bottle
Linear Formula (C6H5)3COH
Quantity 50 g
Beilstein 06,713
Merck Index 15,9912
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in conc. h2so4 and glacial acetic acid,soluble in alcohol,ether and benzene
Formula Weight 260.32
Percent Purity 97%
Physical Form Fine Crystalline Powder
Chemical Name or Material Triphenylmethanol
Show More Show Less
EINECSNumber 200-988-5

RUO – Research Use Only