missing translation for 'onlineSavingsMsg'
Learn More

Triphenylmethanethiol 97.0+%, TCI America™

Catalog No. p-7137127
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T22445G 5 g
T224425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T22445G Supplier TCI America Supplier No. T22445G
Only null left

Chemical Identifiers

CAS 3695-77-0
Molecular Formula C19H16S
Molecular Weight (g/mol) 276.397
MDL Number MFCD00004854
InChI Key JQZIKLPHXXBMCA-UHFFFAOYSA-N
Synonym triphenylmethyl mercaptan, trityl mercaptan, tritylthiol, triphenylmethylmercaptan, methanethiol, triphenyl, tritylmercaptan, triphenyl-methanethiol, alpha,alpha-diphenylbenzenemethanethiol, benzenemethanethiol, alpha,alpha-diphenyl, benzenemethanethiol, .alpha.,.alpha.-diphenyl
PubChem CID 77281
IUPAC Name triphenylmethanethiol
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)S

Specifications

Melting Point 108°C
Color Yellow
Quantity 5 g
Formula Weight 276.40
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Triphenylmethanethiol
RTECSNumber PB5335000
TSCA Yes