missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triphenylmethane 98.0+%, TCI America™
Supplier: TCI America T0515500G
Specifications
| Triphenylmethane | |
| 94°C | |
| 359°C | |
| MFCD00004763 | |
| triphenylmethane, tritane, benzene, 1,1',1-methylidynetris, methane, triphenyl, diphenylmethyl benzene, 1,1',1-methanetriyltribenzene, unii-8o4utw9e17, ccris 5194, triphenyl methane, triphenylmethane? | |
| C1=CC=C(C=C1)C(C1=CC=CC=C1)C1=CC=CC=C1 | |
| 244.34 | |
| CHEBI:76212 | |
| ≥98.0% (GC) |
| 519-73-3 | |
| White-Yellow | |
| C19H16 | |
| 500 g | |
| AAAQKTZKLRYKHR-UHFFFAOYSA-N | |
| (diphenylmethyl)benzene | |
| 10614 | |
| 244.34 | |
| Crystalline Powder |
Chemical Identifiers
| 519-73-3 | |
| 244.34 | |
| AAAQKTZKLRYKHR-UHFFFAOYSA-N | |
| 10614 | |
| (diphenylmethyl)benzene |
| C19H16 | |
| MFCD00004763 | |
| triphenylmethane, tritane, benzene, 1,1',1-methylidynetris, methane, triphenyl, diphenylmethyl benzene, 1,1',1-methanetriyltribenzene, unii-8o4utw9e17, ccris 5194, triphenyl methane, triphenylmethane? | |
| CHEBI:76212 | |
| C1=CC=C(C=C1)C(C1=CC=CC=C1)C1=CC=CC=C1 |
Safety and Handling
TSCA : Yes