missing translation for 'onlineSavingsMsg'
Learn More

Triphenylmethane 98.0+%, TCI America™

Catalog No. T051525G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T051525G 25 g
T0515100G 100 g
T0515500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T051525G Supplier TCI America Supplier No. T051525G

Chemical Identifiers

CAS 519-73-3
Molecular Formula C19H16
Molecular Weight (g/mol) 244.34
MDL Number MFCD00004763
InChI Key AAAQKTZKLRYKHR-UHFFFAOYSA-N
Synonym triphenylmethane, tritane, benzene, 1,1',1-methylidynetris, methane, triphenyl, diphenylmethyl benzene, 1,1',1-methanetriyltribenzene, unii-8o4utw9e17, ccris 5194, triphenyl methane, triphenylmethane?
PubChem CID 10614
ChEBI CHEBI:76212
IUPAC Name (diphenylmethyl)benzene
SMILES C1=CC=C(C=C1)C(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 94°C
Color White-Yellow
Boiling Point 359°C
Quantity 25 g
Formula Weight 244.34
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Triphenylmethane