missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triphenylethylene, 98+%
CAS: 58-72-0 | C20H16 | 256.348 g/mol
$125.34 - $1747.66
Chemical Identifiers
| CAS | 58-72-0 |
|---|---|
| Molecular Formula | C20H16 |
| Molecular Weight (g/mol) | 256.348 |
| MDL Number | MFCD00004765 |
| InChI Key | MKYQPGPNVYRMHI-UHFFFAOYSA-N |
| Synonym | triphenylethylene, triphenylethene, ethylene, triphenyl, benzilidenediphenylmethane, 1,1,2-triphenylethylene, ethene-1,1,2-triyltribenzene, 1,2-diphenylvinyl benzene, 1,2,2-triphenylethylene, benzene, 1,1',1-1-ethenyl-2-ylidene tris, unii-s4zlz1k74b |
| PubChem CID | 6025 |
| ChEBI | CHEBI:35034 |
| IUPAC Name | 1,2-diphenylethenylbenzene |
| SMILES | C1=CC=C(C=C1)C=C(C2=CC=CC=C2)C3=CC=CC=C3 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1324409
|
Thermo Scientific Chemicals
A1324409 |
10 g |
Each for $125.34
|
|
|||||
|
AAA1324418
|
Thermo Scientific Chemicals
A1324418 |
50 g |
Each for $458.97
|
|
|||||
|
AAA1324430
|
Thermo Scientific Chemicals
A1324430 |
250 g |
Each for $1,747.66
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 58-72-0 | |
| 256.348 | |
| MKYQPGPNVYRMHI-UHFFFAOYSA-N | |
| 6025 | |
| 1,2-diphenylethenylbenzene |
| C20H16 | |
| MFCD00004765 | |
| triphenylethylene, triphenylethene, ethylene, triphenyl, benzilidenediphenylmethane, 1,1,2-triphenylethylene, ethene-1,1,2-triyltribenzene, 1,2-diphenylvinyl benzene, 1,2,2-triphenylethylene, benzene, 1,1',1-1-ethenyl-2-ylidene tris, unii-s4zlz1k74b | |
| CHEBI:35034 | |
| C1=CC=C(C=C1)C=C(C2=CC=CC=C2)C3=CC=CC=C3 |
Specifications
| 58-72-0 | |
| 219°C to 221°C (14 mmHg) | |
| MFCD00004765 | |
| 1867462 | |
| MKYQPGPNVYRMHI-UHFFFAOYSA-N | |
| 1,2-diphenylethenylbenzene | |
| 6025 | |
| 256.35 | |
| Triphenylethylene |
| 67°C to 71°C | |
| C20H16 | |
| 10 g | |
| triphenylethylene, triphenylethene, ethylene, triphenyl, benzilidenediphenylmethane, 1,1,2-triphenylethylene, ethene-1,1,2-triyltribenzene, 1,2-diphenylvinyl benzene, 1,2,2-triphenylethylene, benzene, 1,1',1-1-ethenyl-2-ylidene tris, unii-s4zlz1k74b | |
| C1=CC=C(C=C1)C=C(C2=CC=CC=C2)C3=CC=CC=C3 | |
| 256.348 | |
| CHEBI:35034 | |
| ≥98% |
Safety and Handling
P260-P264b-P270-P301+P312-P314-P330-P501c
H302-H373
EINECSNumber : 200-395-1
RTECSNumber : KX4920000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only