Learn More
Triphenylethylene, 98+%
CAS: 58-72-0 | C20H16 | 256.348 g/mol
Supplier: Thermo Scientific Chemicals A1324418
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Triphenylethylene | |
| 67°C to 71°C | |
| C20H16 | |
| 50 g | |
| triphenylethylene, triphenylethene, ethylene, triphenyl, benzilidenediphenylmethane, 1,1,2-triphenylethylene, ethene-1,1,2-triyltribenzene, 1,2-diphenylvinyl benzene, 1,2,2-triphenylethylene, benzene, 1,1',1-1-ethenyl-2-ylidene tris, unii-s4zlz1k74b | |
| C1=CC=C(C=C1)C=C(C2=CC=CC=C2)C3=CC=CC=C3 | |
| 256.348 | |
| CHEBI:35034 | |
| ≥98% |
| 58-72-0 | |
| 219°C to 221°C (14 mmHg) | |
| MFCD00004765 | |
| 1867462 | |
| MKYQPGPNVYRMHI-UHFFFAOYSA-N | |
| 1,2-diphenylethenylbenzene | |
| 6025 | |
| 256.35 |
Chemical Identifiers
| 58-72-0 | |
| 256.348 | |
| MKYQPGPNVYRMHI-UHFFFAOYSA-N | |
| 6025 | |
| 1,2-diphenylethenylbenzene |
| C20H16 | |
| MFCD00004765 | |
| triphenylethylene, triphenylethene, ethylene, triphenyl, benzilidenediphenylmethane, 1,1,2-triphenylethylene, ethene-1,1,2-triyltribenzene, 1,2-diphenylvinyl benzene, 1,2,2-triphenylethylene, benzene, 1,1',1-1-ethenyl-2-ylidene tris, unii-s4zlz1k74b | |
| CHEBI:35034 | |
| C1=CC=C(C=C1)C=C(C2=CC=CC=C2)C3=CC=CC=C3 |
Safety and Handling
P260-P264b-P270-P301+P312-P314-P330-P501c
H302-H373
EINECSNumber : 200-395-1
RTECSNumber : KX4920000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only