missing translation for 'onlineSavingsMsg'
Learn More

Triphenylene 96.0+%, TCI America™

Supplier:  TCI America T05131G

 View more versions of this product

Catalog No. T05131G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Triphenylene
199°C
C18H12
1 g
SLGBZMMZGDRARJ-UHFFFAOYSA-N
triphenylene
9170
228.29
Crystalline Powder
217-59-4
425°C
MFCD00001108
9,10-benzophenanthrene, isochrysene, 9,10-benzphenanthrene, benzo l phenanthrene, 1,2,3,4-dibenznaphthalene, benzophenanthrene, 1,2:3,4-dibenznaphthalene, ccris 1301, triphenylen-1-yl
C1=CC=C2C(=C1)C1=CC=CC=C1C1=CC=CC=C21
228.29
CHEBI:33080
≥96.0% (GC)

Chemical Identifiers

217-59-4
228.29
SLGBZMMZGDRARJ-UHFFFAOYSA-N
9170
triphenylene
C18H12
MFCD00001108
9,10-benzophenanthrene, isochrysene, 9,10-benzphenanthrene, benzo l phenanthrene, 1,2,3,4-dibenznaphthalene, benzophenanthrene, 1,2:3,4-dibenznaphthalene, ccris 1301, triphenylen-1-yl
CHEBI:33080
C1=CC=C2C(=C1)C1=CC=CC=C1C1=CC=CC=C21
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : YK2925000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions