missing translation for 'onlineSavingsMsg'
Learn More

Triphenylene 96.0+%, TCI America™

Catalog No. T0513100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0513100MG 100 mg
T05131G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T0513100MG Supplier TCI America Supplier No. T0513100MG

Chemical Identifiers

CAS 217-59-4
Molecular Formula C18H12
Molecular Weight (g/mol) 228.29
MDL Number MFCD00001108
InChI Key SLGBZMMZGDRARJ-UHFFFAOYSA-N
Synonym 9,10-benzophenanthrene, isochrysene, 9,10-benzphenanthrene, benzo l phenanthrene, 1,2,3,4-dibenznaphthalene, benzophenanthrene, 1,2:3,4-dibenznaphthalene, ccris 1301, triphenylen-1-yl
PubChem CID 9170
ChEBI CHEBI:33080
IUPAC Name triphenylene
SMILES C1=CC=C2C(=C1)C1=CC=CC=C1C1=CC=CC=C21

Specifications

Melting Point 199°C
Boiling Point 425°C
Quantity 100 mg
Formula Weight 228.29
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Triphenylene
RTECSNumber YK2925000
TSCA No