missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triphenylene (purified by sublimation) 98.0+%, TCI America™
Supplier: TCI America T32671G
Specifications
| Triphenylene (purified by sublimation) | |
| 199°C | |
| C18H12 | |
| 1 g | |
| SLGBZMMZGDRARJ-UHFFFAOYSA-N | |
| triphenylene | |
| 9170 | |
| 228.29 | |
| Crystalline Powder |
| 217-59-4 | |
| 425°C | |
| MFCD00001108 | |
| 9,10-benzophenanthrene, isochrysene, 9,10-benzphenanthrene, benzo l phenanthrene, 1,2,3,4-dibenznaphthalene, benzophenanthrene, 1,2:3,4-dibenznaphthalene, ccris 1301, triphenylen-1-yl | |
| C1=CC=C2C(=C1)C1=CC=CC=C1C1=CC=CC=C21 | |
| 228.29 | |
| CHEBI:33080 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 217-59-4 | |
| 228.29 | |
| SLGBZMMZGDRARJ-UHFFFAOYSA-N | |
| 9170 | |
| triphenylene |
| C18H12 | |
| MFCD00001108 | |
| 9,10-benzophenanthrene, isochrysene, 9,10-benzphenanthrene, benzo l phenanthrene, 1,2,3,4-dibenznaphthalene, benzophenanthrene, 1,2:3,4-dibenznaphthalene, ccris 1301, triphenylen-1-yl | |
| CHEBI:33080 | |
| C1=CC=C2C(=C1)C1=CC=CC=C1C1=CC=CC=C21 |
Safety and Handling
RTECSNumber : YK2925000
TSCA : No