missing translation for 'onlineSavingsMsg'
Learn More

Triphenylcarbenium tetrafluoroborate, 97%

CAS: 341-02-6 | C19H15·BF4 | 330.13 g/mol

$80.96 - $80.96

Chemical Identifiers

CAS 341-02-6
MDL Number MFCD00013120
InChI Key VQXBOEYKSVVPSP-UHFFFAOYSA-N
Synonym triphenylcarbenium tetrafluoroborate, trityl tetrafluoroborate, tritylium tetrafluoroborate, triphenylmethylium tetrafluoroborate, diphenylmethylbenzene tetrafluoroborate, trityl fluoroborate, triphenylmethyl fluoroborate, triphenylcarbonium tetrafluoroborate, methylium, triphenyl-, tetrafluoroborate 1-, methylium, triphenyl-, tetrafluoroborate 1-1:1
PubChem CID 2723955
IUPAC Name diphenylmethylbenzene;tetrafluoroborate
SMILES [B-](F)(F)(F)F.C1=CC=C(C=C1)[C+](C2=CC=CC=C2)C3=CC=CC=C3
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC172950050
SDS View Documents
Thermo Scientific Chemicals
172950050
5 g
Each for $80.96
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

341-02-6
VQXBOEYKSVVPSP-UHFFFAOYSA-N
2723955
[B-](F)(F)(F)F.C1=CC=C(C=C1)[C+](C2=CC=CC=C2)C3=CC=CC=C3
MFCD00013120
triphenylcarbenium tetrafluoroborate, trityl tetrafluoroborate, tritylium tetrafluoroborate, triphenylmethylium tetrafluoroborate, diphenylmethylbenzene tetrafluoroborate, trityl fluoroborate, triphenylmethyl fluoroborate, triphenylcarbonium tetrafluoroborate, methylium, triphenyl-, tetrafluoroborate 1-, methylium, triphenyl-, tetrafluoroborate 1-1:1
diphenylmethylbenzene;tetrafluoroborate

Specifications

341-02-6
Green to Ochre-Yellow
97%
C19H15·BF4
MFCD00013120
triphenylcarbenium tetrafluoroborate, trityl tetrafluoroborate, tritylium tetrafluoroborate, triphenylmethylium tetrafluoroborate, diphenylmethylbenzene tetrafluoroborate, trityl fluoroborate, triphenylmethyl fluoroborate, triphenylcarbonium tetrafluoroborate, methylium, triphenyl-, tetrafluoroborate 1-, methylium, triphenyl-, tetrafluoroborate 1-1:1
VQXBOEYKSVVPSP-UHFFFAOYSA-N
diphenylmethylbenzene;tetrafluoroborate
2723955
97%
Triphenylcarbenium tetrafluoroborate
205°C to 215°C
Authentic
Plastic bottle
(C6H5)3CBF4
5 g
Solubility in water: insoluble. Other solubilities: soluble in methanol
[B-](F)(F)(F)F.C1=CC=C(C=C1)[C+](C2=CC=CC=C2)C3=CC=CC=C3
330.13
330.13
Crystalline Powder, Crystals or Chunks
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Reacts violently with water.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 206-433-3

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only