missing translation for 'onlineSavingsMsg'
Learn More

Triphenylcarbenium tetrafluoroborate, 97%

Catalog No. AAA1294909
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1294909 10 g
AAA1294918 50 g
AAA1294930 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1294909 Supplier Thermo Scientific Chemicals Supplier No. A1294909

CAS: 341-02-6 | C19H15BF4 | 330.133 g/mol

It is applied in the abstraction of the-H atom of alcohols and their O-substituted derivatives results in oxidation to the ketones and in the oxidation of TMS ethers to ketones, secondary alcohols to ketones, and secondary diols to ?-hydroxy ketones. It is applied as a reactant involved in the synthesis of substituted dihydroazulene photo switches, carbene-based Lewis pairs for hydrogen activation, 1,3-Dipolar cycloaddition reactions for preparation of α-amino-β-hydroxy esters, oxidation of allenic compounds, Mukaiyama aldol addition reactions and ionic hydrogenation as a counteranion and ligand source.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied in the abstraction of the ɑ-H atom of alcohols and their O-substituted derivatives results in oxidation to the ketones and in the oxidation of TMS ethers to ketones, secondary alcohols to ketones, and secondary diols to ɑ-hydroxy ketones. It is applied as a reactant involved in the synthesis of substituted dihydroazulene photo switches, carbene-based Lewis pairs for hydrogen activation, 1,3-Dipolar cycloaddition reactions for preparation of α-amino-β-hydroxy esters, oxidation of allenic compounds, Mukaiyama aldol addition reactions and ionic hydrogenation as a counteranion and ligand source.

Solubility
Insoluble in water. Soluble in methanol.

Notes
Moisture sensitive. Store away from bases, water/moisture, heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Keep it refrigerated.

Chemical Identifiers

CAS 341-02-6
Molecular Formula C19H15BF4
Molecular Weight (g/mol) 330.133
MDL Number MFCD00013120
InChI Key VQXBOEYKSVVPSP-UHFFFAOYSA-N
Synonym triphenylcarbenium tetrafluoroborate, trityl tetrafluoroborate, tritylium tetrafluoroborate, triphenylmethylium tetrafluoroborate, diphenylmethylbenzene tetrafluoroborate, trityl fluoroborate, triphenylmethyl fluoroborate, triphenylcarbonium tetrafluoroborate, methylium, triphenyl-, tetrafluoroborate 1-, methylium, triphenyl-, tetrafluoroborate 1-1:1
PubChem CID 2723955
IUPAC Name diphenylmethylbenzene;tetrafluoroborate
SMILES [B-](F)(F)(F)F.C1=CC=C(C=C1)[C+](C2=CC=CC=C2)C3=CC=CC=C3

Specifications

Melting Point ∼207°C (decomposition)
Odor Odorless
Linear Formula (C6H5)3CBF4
Quantity 10 g
UN Number UN3261
Beilstein 5085649
Sensitivity Moisture sensitive
Solubility Information Insoluble in water. Soluble in methanol.
Formula Weight 330.13
Percent Purity 98%
Chemical Name or Material Triphenylcarbenium tetrafluoroborate
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 206-433-3
TSCA Yes
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only