missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triphenylborane - Triphenylphosphine Complex 97.0+%, TCI America™
$111.84 - $331.81
Chemical Identifiers
| CAS | 3053-68-7 |
|---|---|
| Molecular Formula | C36H30BP |
| Molecular Weight (g/mol) | 504.42 |
| MDL Number | MFCD01631317 |
| InChI Key | QLAGHGSFXJZWKY-UHFFFAOYSA-N |
| PubChem CID | 11081676 |
| IUPAC Name | triphenylborane;triphenylphosphane |
| SMILES | B(C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3 |
Chemical Identifiers
| 3053-68-7 | |
| 504.42 | |
| QLAGHGSFXJZWKY-UHFFFAOYSA-N | |
| triphenylborane;triphenylphosphane |
| C36H30BP | |
| MFCD01631317 | |
| 11081676 | |
| B(C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3 |
Specifications
| 3053-68-7 | |
| C36H30BP | |
| 5 g | |
| B(C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3 | |
| 504.42 | |
| 504.42 | |
| Crystalline Powder |
| White-Yellow | |
| MFCD01631317 | |
| QLAGHGSFXJZWKY-UHFFFAOYSA-N | |
| triphenylborane;triphenylphosphane | |
| 11081676 | |
| ≥97.0% (T) | |
| Triphenylborane - Triphenylphosphine Complex |
Safety and Handling
TSCA : No