missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triphenylborane - Triphenylphosphine Complex 97.0+%, TCI America™
Specifications
Specifications
| Color | White-Yellow |
| Quantity | 5 g |
| Formula Weight | 504.42 |
| Percent Purity | ≥97.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Triphenylborane - Triphenylphosphine Complex |
Safety and Handling
| TSCA | No |